Learn More
MilliporeSigma™ Certipur™ Refractive Index Standard Kits
Refractive index standards Certipur™ according to Ph Eur.
Supplier:  MilliporeSigma™ 1089620001
 View more versions of this product
Catalog No. M1089620001
Only null left
Explore available promotions
Description
Description
These standards are for the calibration of refractometers. We offer different refractive index standards in order to perform calibration in the same range as the final measurement.
  • Stable solutions with minimum shelf life of 3 years
  • Ready-to-use solutions
  • Each standard kit is packed with a certificate of analysis
  • Applicable at 20°C and 25°C measuring temperature
  • According to Ph Eur. 2.2.6
Specifications
Specifications
Traceable to SRM from NIST and PTB n (20°C)=1,3915 Certipur
2,2,4-Trimethyl-pentane/Water
Product Suggestions
Product Suggestions