Learn More
Mettler Toledo™ Certified Temperature Kits
Catalog No. 01915001
Change view
Click to view available options
For Use With (Equipment):
HE73, HE53 Moisture Analyzer
HX204, HS153, HC103 Moisture Analyzer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01915001 HE73, HE53 Moisture Analyzer
01915002 HX204, HS153, HC103 Moisture Analyzer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01915001 Supplier Mettler Toledo™ Supplier No. 30134141
Quick and easy temperature testing, for use with Mettler Toledo™ Moisture Analyzers
  • Quick and easy temperature testing
  • Adjusts instrument at the selected operating temperature to ensure reliable results
  • For use with Mettler Toledo™ Moisture Analyzers
Specifications
For Use With (Equipment) HE73, HE53 Moisture Analyzer
Type Temperature Kit