Learn More
Certified Screw Top Vial Kits with Pico Pure Plus+™ Caps, Sold by MilliporeSigma™ Supelco™
Developed for the LC/MS market. It is a one-piece septumless closure system and does not have the contamination issues that is found is other closures.
$118.00
Specifications
Volume (Metric) 2 mL
No. per Pack 100
Closure Size 9 mm
View More Specs
Products 1
Catalog Number Mfr. No. Marking Spot Color Price Quantity & Availability  
Catalog Number Mfr. No. Marking Spot Color Price Quantity & Availability  
111011268
View Documents Promotion Details
N/A Yes White, Clear
Each for $118.00
Only null left
Description
Description
  • The impurities of this cap is well below the picogram range of purity for extractables.
  • This closure seals as well as a vial and cap with a PTFE/silicone septa
  • This product will puncture with far less force than what is needed to puncture a PTFE/silicone septa.
  • The inside diameter of vials vary with the manufacture.
  • The following insert part numbers are compatible with these large opening vials.. 29435-U PK100, Certified 200 muL glass inserts, mandrel precision point, 6 x 29 mm, with bottom spring. 29436-U PK100, Certified 250 muL glass insert, conical bottom, 6 x 31 mm. 29437-U PK100, Certified 300 muL glass inserts, conical bottom, 6 x 30 mm. 29441-U PK100, Certified 350 muL glass inserts, flat bottom, 6 x 31 mm
Specifications
Specifications
2 mL
100
9 mm
Videos
SDS
Documents
Documents
Pico Pure Plus+ is a trademark of AR Corp.