missing translation for 'onlineSavingsMsg'
Learn More

Cerium oxide, 20% in H{2}O, colloidal dispersion

Widely applied as a solid electrolyte in fuel cells. Also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. | CAS: 1306-38-3 | CeO2 | 172.114 g/mol

Supplier:  Thermo Scientific Chemicals 40125A3

 View more versions of this product

Catalog No. AA40125A3


Only null left
Explore available promotions

Description

Description

Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium oxide
Particles in Liquid
CeO2
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CETPSERCERDGAM-UHFFFAOYSA-N
dioxocerium
73963
172.12
Plastic bottle
1.22
1306-38-3
2 kg
MFCD00010933
Immiscible with water.
O=[Ce]=O
172.114
CHEBI:79089
20% in H2O
1.22

Chemical Identifiers

1306-38-3
172.114
CETPSERCERDGAM-UHFFFAOYSA-N
73963
dioxocerium
CeO2
MFCD00010933
ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
CHEBI:79089
O=[Ce]=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-150-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only