Learn More
Cerium(IV) oxide, REacton™, 99.99% (REO)
CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Supplier: Thermo Scientific Chemicals 01132722
Description
Cerium(IV) oxide is used in ceramics and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cerium(IV) oxide | |
| 1306-38-3 | |
| Powder | |
| 99.99% | |
| MFCD00010933 | |
| Insoluble in water. | |
| O=[Ce]=O | |
| 172.114 | |
| CHEBI:79089 | |
| Odorless |
| 2600°C | |
| Yellow | |
| 100 g | |
| CeO2 | |
| ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
| CETPSERCERDGAM-UHFFFAOYSA-N | |
| dioxocerium | |
| 73963 | |
| 172.12 | |
| 7.132 |
Chemical Identifiers
| 1306-38-3 | |
| 172.114 | |
| CETPSERCERDGAM-UHFFFAOYSA-N | |
| 73963 | |
| dioxocerium |
| CeO2 | |
| MFCD00010933 | |
| ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline | |
| CHEBI:79089 | |
| O=[Ce]=O |
Safety and Handling
H302 : Harmful if swallowed.
EINECSNumber : 215-150-4
RTECSNumber : FK6310000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only