Learn More
Cerium(IV) oxide, REacton™, 99.99% (REO)
Description
Cerium(IV) oxide is used in ceramics and to polish glass and stones. It is applied as a solid electrolyte in fuel cells. It can also be employed as a co-catalyst in a number of reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2600°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.99% |
| Solubility Information | Insoluble in water. |
| Formula Weight | 172.12 |
| Odor | Odorless |
| Density | 7.132 g/cm3 at 20°C |
| Chemical Name or Material | Cerium(IV) oxide |
Safety and Handling
| Hazard Category | H302 : Harmful if swallowed. |
| EINECSNumber | 215-150-4 |
| RTECSNumber | FK6310000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only