missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) sulfide, 99.9% (metals basis)

Catalog No. AA4185606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4185606 5 g
AA4185614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4185606 Supplier Thermo Scientific Chemicals Supplier No. 04185606

CAS: 12014-93-6 | Ce2S3 | 376.41 g/mol

Used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12014-93-6
Molecular Formula Ce2S3
Molecular Weight (g/mol) 376.41
MDL Number MFCD00016007
InChI Key MMXSKTNPRXHINM-UHFFFAOYSA-N
Synonym cerium sulfide, dicerium trisulphide, ceric sulfide, cerium 3+ trisulfide, dicerium 3+ trisulfide, dicerium 3+ trisulfanediide
PubChem CID 159394
SMILES [S--].[S--].[S--].[Ce+3].[Ce+3]

Specifications

Melting Point 2100°C
Color Red to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name cerium(3+);trisulfide
Formula Weight 376.43
Odor Rotten eggs
Appearance Red-brown powder
Density 5.02 g/cm3 at 20°C
Chemical Name or Material Cerium(III) sulfide
Show More Show Less
Hazard Category H261-H315-H319-H335
Precautionary Statement P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 4.3, Packaging Group: III
EINECSNumber 234-603-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only