accessibility menu, dialog, popup

UserName

CAS RN 12014-93-6

IUPAC Name: dicerium(3+) trisulfanediide
Synonyms: dicerium(3+) trisulfanediide
Molecular Weight (g/mol): 376.41
Molecular Formula: Ce2S3
InChi Key: MMXSKTNPRXHINM-UHFFFAOYSA-N
SMILES: [S--].[S--].[S--].[Ce+3].[Ce+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results