Learn More
Cerium(III) nitrate hexahydrate, 99.5%
CAS: 10294-41-4 | CeH12N3O15 | 434.22 g/mol
Supplier: Thermo Scientific Chemicals 218690025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cerium(III) nitrate hexahydrate | |
| 10294-41-4 | |
| Crystalline Powder or Agglomerating Crystals | |
| 99.4% min. (CeO2/TREO basis) | |
| Ce(NO3)3·6H2O | |
| 16,67 | |
| cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f | |
| QQZMWMKOWKGPQY-UHFFFAOYSA-N | |
| cerium(3+) hexahydrate trinitrate | |
| 16211466 | |
| 99.5% |
| 99.5% | |
| Colorless to White | |
| 2.5 kg | |
| CeH12N3O15 | |
| MFCD00149631 | |
| 10,1959 | |
| Solubility in water: 1754g/L (25°C). Other solubilities: soluble in alcohol and acetone | |
| O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 434.22 | |
| 434.22 | |
| Plastic bottle |
Chemical Identifiers
| 10294-41-4 | |
| 434.22 | |
| QQZMWMKOWKGPQY-UHFFFAOYSA-N | |
| 16211466 |
| CeH12N3O15 | |
| MFCD00149631 | |
| cerium iii nitrate hexahydrate, cerium nitrate hexahydrate, cerous nitrate hexahydrate, unii-fl1r38jebb, fl1r38jebb, nitric acid cerium salt, cerium iii nitratehexahydrate, ce.3no3.6h2o, ksc494m5f | |
| O.O.O.O.O.O.[Ce+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
May intensify fire; oxidizer.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take any precaution to avoid mixing with combustibles.
In case of major
GHS Signal Word: Danger
RTECSNumber : FK6300000
RUO – Research Use Only