missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) Chloride Anhydrous 98.0+%, TCI America™
Supplier: TCI America C205825G
Specifications
| Cerium(III) Chloride Anhydrous | |
| White-Yellow | |
| Crystalline Powder | |
| CeCl3 | |
| VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
| cerium(3+);trichloride | |
| 24636 | |
| 246.47 |
| 722°C | |
| 7790-86-5 | |
| 25 g | |
| cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
| [Cl-].[Cl-].[Cl-].[Ce+3] | |
| 246.466 | |
| CHEBI:35458 | |
| ≥98.0% (T) |
Chemical Identifiers
| 7790-86-5 | |
| 246.466 | |
| cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride | |
| CHEBI:35458 | |
| [Cl-].[Cl-].[Cl-].[Ce+3] |
| CeCl3 | |
| VYLVYHXQOHJDJL-UHFFFAOYSA-K | |
| 24636 | |
| cerium(3+);trichloride |
Safety and Handling
EINECSNumber : (1)-0622
RTECSNumber : FK5075000
TSCA : Yes