accessibility menu, dialog, popup

UserName

CAS RN 7790-86-5

IUPAC Name: cerium(3+) trichloride
Synonyms: cerium(III) chloride
Molecular Weight (g/mol): 246.47
Molecular Formula: CeCl3
InChi Key: VYLVYHXQOHJDJL-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Ce+3]
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)