missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) Chloride Anhydrous 98.0+%, TCI America™

Catalog No. C20585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20585G 5 g
C205825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20585G Supplier TCI America Supplier No. C20585G

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.466
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Melting Point 722°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 246.47
Percent Purity ≥98.0% (T)
Chemical Name or Material Cerium(III) Chloride Anhydrous
EINECSNumber (1)-0622
RTECSNumber FK5075000
TSCA Yes