missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride, 99.9%, pure, anhydrous

Catalog No. p-3734717
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369750050 5 g Glass bottle
AC369750250 25 g Glass bottle
AC369751000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC369750050 Supplier Thermo Scientific Chemicals Supplier No. 369750050
Only null left

CAS: 7790-86-5 | CeCl3 | 246.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.48
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Boiling Point 1727.0°C
Melting Point 848.0°C
Color White to Gray
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.90%
Linear Formula CeCl3
Merck Index 15, 1996
Solubility Information Solubility in water: soluble
Formula Weight 246.48
Percent Purity 99.9%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Cerium(III) chloride, Anhdrous
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 232-227-8

RUO – Research Use Only