missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide hydrate, 99.998% (metals basis)

Catalog No. p-7059334
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4443209 10 g
AA4443218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4443209 Supplier Thermo Scientific Chemicals Supplier No. 04443209
Only null left

CAS: 14457-87-5 | Br3Ce | 379.83 g/mol

Cerium(III) bromide is used in the following ways, CeBr3-doped lanthanum bromide single crystals are known to exhibit superior scintillation properties for applications in the security, medical imaging, and geophysics detectors.Undoped single crystals of CeBr3 have shown promise as a γ-ray scintillation detector in nuclear non-proliferation testing, medical imaging, environmental remediation, and oil exploration

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14457-87-5
Molecular Formula Br3Ce
Molecular Weight (g/mol) 379.83
MDL Number MFCD00016004
InChI Key MOOUSOJAOQPDEH-UHFFFAOYSA-K
IUPAC Name cerium(3+) tribromide
SMILES [Br-].[Br-].[Br-].[Ce+3]

Specifications

Color Orange
Physical Form Crystalline
Quantity 10 g
Merck Index 14,1995
Solubility Information Soluble in water.
Formula Weight 379.83 (Anhydrous)
Percent Purity 99.998%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Cerium (III) bromide hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 238-447-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only