CAS RN 14457-87-5
IUPAC Name:
cerium(3+) tribromide
Synonyms:
cerium(III) bromide
Molecular Weight (g/mol):
379.83
Molecular Formula:
Br3Ce
InChi Key:
MOOUSOJAOQPDEH-UHFFFAOYSA-K
SMILES:
[Br-].[Br-].[Br-].[Ce+3]
1
–
3
of
3
results
1
Cerium(III) bromide hydrate, 99.998% (metals basis)
CAS: 14457-87-5 Molecular Formula: Br3Ce Molecular Weight (g/mol): 379.83 MDL Number: MFCD00016004 InChI Key: MOOUSOJAOQPDEH-UHFFFAOYSA-K IUPAC Name: cerium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Ce+3]
2
Cerium(III) bromide, ultra dry, 99.99% (metals basis)
CAS: 14457-87-5 Molecular Formula: Br3Ce Molecular Weight (g/mol): 379.83 MDL Number: MFCD00016004 InChI Key: MOOUSOJAOQPDEH-UHFFFAOYSA-K IUPAC Name: cerium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Ce+3]
3
Cerium(III) bromide hydrate, 99%
CAS: 14457-87-5 Molecular Formula: Br3Ce Molecular Weight (g/mol): 379.83 MDL Number: MFCD00016004 InChI Key: MOOUSOJAOQPDEH-UHFFFAOYSA-K IUPAC Name: cerium(3+) tribromide SMILES: [Br-].[Br-].[Br-].[Ce+3]