missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Zopiclone-N-oxide Solution, 100 μg/mL (Methanol with 1% 1 M HCl), Sold by MilliporeSigma™ Supelco™
Description
Zopiclone N-oxide is a primary, active metabolite of zopiclone in urine as well as a major pharmaceutical impurity of zopiclone known as eszopiclone impurity A. The parent drug, zopiclone, is a non-benzodiazepine hypnotic sold under the trade names Imovane™and ZImovane™and is used in the treatment of insomnia.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 100 μg/mL (methanol with 1% 1 M HCl) |
| Linear Formula | C17H17ClN6O4 |
| MDL Number | MFCD00871938 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol |
| Quantity | 1 mL |
| Formula Weight | 404.81 |
CERILLIANT is a registered trademark of Cerilliant Corporation