missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Zolpidem phenyl-4-carboxylic acid Solution, 500 μg/mL in Acetonitrile:Water (1:1), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002191
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002191 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002191 Supplier Merck Emd Millipore Supplier No. Z0071ML

Certified reference material

Zolpidem Phenyl-4-carboxylic acid is the major urinary metabolite of zolpidem, accounting for 51% of an administered dose. Literature references indicate the metabolite can be found in urine for 2-3 days following ingestion of a single therapeutic dose of zolpidem. Zolpidem, marketed as Ambien™ or Stilnox™, is a nonbenzodiazepine used for the short-term treatment of insomnia.

Specifications

CAS 75-05-8, 109461-65-6
MDL Number MFCD18252339
UN Number UN 1648 3 / PGII
Concentration 500 μg/mL in acetonitrile:water (1:1)
Packaging Ampule
Quantity 1 mL
Flash Point 13°C (55.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation