missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Zolpidem Phenyl-4-carboxylic acid-D4 Solution, 100 μg/mL in Acetonitrile:Water, Sold by MilliporeSigma™ Supelco™
Description
A stable-labeled internal standard suitable for quantitation of Zolpidem Phenyl-4-carboxylic acid levels in urine or other matrices by LC/MS or GC/MS for testing applications in clinical toxicology, forensic analysis, or urine drug testing. Zolpidem Phenyl-4-carboxylic acid is the major urinary metabolite of zolpidem, accounting for 51% of an administered dose. Literature references indicate the metabolite can be found in urine for 2-3 days following ingestion of a single therapeutic dose of zolpidem. Zolpidem, marketed as Ambien or Stilnox, is a nonbenzodiazepine used for the short-term treatment of insomnia.
Specifications
Specifications
| CAS | 75-05-8 |
| Linear Formula | C19H15D4N3O3 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile:water |
| Formula Weight | 341.40 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 13°C (55.4°F) |
CERILLIANT is a registered trademark of Cerilliant Corporation