missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Voriconazole Solution, 2.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
Voriconazole, sold under the trade name Vfend™, is a triazole antifungal drug prescribed for treatment of serious, invasive fungal infections. This certified Snap-N-Spike™ Solution is suitable as starting material for linearity standards, calibrators, or controls for therapeutic drug monitoring or clinical and diagnostic testing of voriconazole in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.
Specifications
Specifications
| CAS | 67-56-1, 137234-62-9 |
| Linear Formula | C16H14F3N5O |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 2.0 mg/mL in methanol |
| Formula Weight | 349.31 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation