missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Vitamin K1 Solution, 10 μg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™
Description
Vitamin K1 (phylloquinone) is a fat-soluble vitamin that serves as a cofactor for vitamin K-dependent (VKD) proteins and enzymes which activate blood-clotting factors. Vitamin K1 is critical to many physiological processes including bone metabolism, arterial calcification, growth control, and signal transduction.
Specifications
Specifications
| CAS | 64-17-5, 84-80-0 |
| Melting Point | -143.99°C |
| Linear Formula | C31H46O2 |
| MDL Number | MFCD00214063 |
| UN Number | UN 1170 - class 3 - PG 2 - Ethanol, solution |
| Concentration | 10 μg/mL in ethanol |
| Formula Weight | 450.70 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 14°C (57.2°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation