missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Vitamin D3 Solution, 1 mg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002149
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002149 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002149 Supplier Merck Emd Millipore Supplier No. V0251ML

Certified reference material

Vitamin D3 or cholecalciferol is a form of vitamin D produced naturally by the skin when exposed to ultraviolet light. Vitamin D3 is also manufactured industrially to fortify foods or for use in vitamin supplements.

Specifications

CAS 64-17-5, 67-97-0
Linear Formula C27H44O
MDL Number MFCD00078131
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 1 mg/mL in ethanol
Synonym Vitamin D3 solution
Formula Weight 384.64
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F)

CERILLIANT is a registered trademark of Cerilliant Corporation