missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Vitamin D2 Solution, 1 mg/mL in Ethanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002148
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002148 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002148 Supplier Merck Emd Millipore Supplier No. V0241ML
Only null left

Certified reference material

Vitamin D2 is a form of vitamin D also known as ergocalciferol. Marketed under trade names including Deltalin, Drisdol™, and Calcidol™, vitamin D2 is often used as a vitamin D supplement for individuals with vitamin D defficiency.

Specifications

CAS 64-17-5, 50-14-6
Melting Point -144.0°C
Linear Formula C28H44O
MDL Number MFCD00166988
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 1 mg/mL in ethanol
Synonym Vitamin D2 solution
Formula Weight 396.65
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F)
Show More Show Less

CERILLIANT is a registered trademark of Cerilliant Corporation