missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Valeryl fentanyl Hydrochloride Solution, 100 μg/mL in Methanol (as a free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002168
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002168 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002168 Supplier Merck Emd Millipore Supplier No. V0480.5ML

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for valeryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. Valeryl fentanyl is a designer drug and fentanyl analog. In March, 2017, Chinese officials banned manufacturing and sale of valeryl fentanyl in China.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Linear Formula C24H32N2O · HCl
Concentration 100 μg/mL in methanol (as a free base)
MDL Number MFCD30718323
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 400.98

CERILLIANT is a registered trademark of Cerilliant Corporation