missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Valeryl fentanyl-D5 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002178
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002178 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002178 Supplier Merck Emd Millipore Supplier No. V0680.5ML

Certified reference material

Valeryl fentanyl is a designer drug and fentanyl analog. In March 2017, Chinese officials banned manufacturing and sale of valeryl fentanyl in China. This stable-labeled internal standard is suitable for quantification of valeryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, or forensic analysis applications. This Certified Spiking Solution is supplied in a convenient US DEA-exempt solution format.

Specifications

Packaging Ampule
CAS 67-56-1
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C24H27D5N2O · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 406.02

CERILLIANT is a registered trademark of Cerilliant Corporation