missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Trinitroglycerin Solution, 1000 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore T0021ML
Description
Trinitroglycerin, also commonly known as nitroglycerin, is a pharmaceutical vasodilator used to treat heart conditions such as angina and chronic heart failure. This explosive compound is also an ingredient in dynamite and gunpowder. This Certified Spiking Solution™ is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology.Specifications
| 75-05-8,55-63-0 | |
| MFCD00171649 | |
| 1000 μg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
| C3H5N3O9 | |
| UN 1648 3 / PGII | |
| 227.09 | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation