Learn More
CERILLIANT™ (-)-trans-Delta9-THC Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore T0051ML
Catalog No. 01002030
Only null left
Explore available promotions
Description
Description
(-)-δ9-THC is the main active constituent and the primary psychoactive cannabinoid of cannabis. Besides persistent illicit use this drug is suitable as treatment for anorexia and nausea and is sold under the trade name Marinol™. This Certified Spiking Solution is suitable as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.
Specifications
Specifications
67-56-1,1972-08-3
MFCD00083207
1.0 mg/mL in methanol
314.46
1 mL
C21H30O2
UN 1230 - class 3 - PG 2 - Methanol, solution
Dronabinol solution
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation