missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Tetramethylenedisulfotetramine Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 75-05-8, 80-12-6 |
| Linear Formula | C4H8N4O4S2 |
| MDL Number | MFCD00221771 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 240.26 |
| Packaging | Ampule |
| Quantity | 1.2 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation