missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Teriflunomide Solution, 1 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002092
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002092 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002092 Supplier Merck Emd Millipore Supplier No. T1221ML
Only null left

Certified reference material

Teriflunomide, marketed under the trade name Aubagio, is the active metabolite of leflunomide, an immunosuppressive disease-modifying antirheumatic drug, used in active moderate-to-severe rheumatoid arthritis and psoriatic arthritis. An analytical reference standard for use in LC/MS or GC/MS applications including clinical and diagnostic testing such as therapeutic drug monitoring assays.

Specifications

CAS 75-05-8, 163451-81-8
Linear Formula C12H9F3N2O2
UN Number UN 1648 3 / PGII
Concentration 1 mg/mL in acetonitrile
Formula Weight 270.21
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation