Learn More
CERILLIANT™ Teriflunomide-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore T1231ML
Catalog No. 01002093
Only null left
Explore available promotions
Description
Description
This stable-labeled internal standard of teriflunomide is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays. Teriflunomide, also known as Aubagio, is the active metabolite of leflunomide, an immunosuppressive disease-modifying antirheumatic drug, used in active moderate-to-severe rheumatoid arthritis and psoriatic arthritis.
Specifications
Specifications
75-05-8 , 1185240-22-5
UN 1648 3 / PGII
274.23
1 mL
C12H5D4F3N2O2
100 μg/mL in acetonitrile
Ampule
2°C (35.6°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation