missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Sufentanil-D5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001942
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001942 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001942 Supplier Merck Emd Millipore Supplier No. S0181ML
Only null left

Certified reference material

Sufentanil is a short acting synthetic opioid analgesic used as a painkiller during medical operations. This stable-labeled internal standard is suitable for quantitation of sufentanil levels in urine, serum, or plasma by LC/MS or GC/MS for clinical toxicology, pain prescription monitoring, forensic analysis, or isotope dilution methods. Sufentanil is sold under the trade name Sufenta.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol
Linear Formula C22D5H25N2O2S
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 391.58

CERILLIANT is a registered trademark of Cerilliant Corporation