missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Sufentanil Citrate Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001937
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001937 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001937 Supplier Merck Emd Millipore Supplier No. S0081ML

Certified reference material

Sufentanil is a short acting synthetic opioid analgesic most commonly used for pain relief during medical operations. This certified solution standard is applicable to use in clinical toxicology, pain prescription monitoring, or forensic analysis by LC-MS/MS or GC/MS. Sufentanil is sold under the trade name Sufenta.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C22H30N2O2S · C6H8O7
MDL Number MFCD00072116
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 578.67

CERILLIANT is a registered trademark of Cerilliant Corporation