missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Stanozolol Solution, 1.0 mg/mL, Sold by MilliporeSigma™ Supelco™

Catalog No. 01002022
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002022 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002022 Supplier Merck Emd Millipore Supplier No. S9061ML

Certified reference material

An analytical reference standard applicable for use in sports testing, urine drug testing, or forensic analysis by LC-MS/MS or GC/MS. Stanozolol is used as a treatment for osteoporosis and steroid-defficiency, but has also been abused in sports as a performance-enhancing drug.

Specifications

CAS 110-71-4, 10418-03-8
Linear Formula C21H32N2O
UN Number UN 2252 - class 3 - PG 2 - 1,2-Dimethoxyethane solution
Concentration 1.0 mg/mL
Formula Weight 328.49
Packaging Ampule
Quantity 1 mL
Flash Point -2°C (28.4°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation