missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Spice Cannabinoid Mix 3 Solution, 100 μg/mL each component in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A certified multi-component solution mix of Spice cannabinoids RCS-4, RCS-8, JWH-015, JWH-203, JWH-210, and AM2233. This mix is suitable as starting material for calibrators and controls in Spice cannabinoid testing methods by GC/MS or LC/MS for urine drug testing, forensic analysis, and clinical toxicology applications.
Specifications
Specifications
| CAS | 75-05-8 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL each component in acetonitrile |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) |
CERILLIANT is a registered trademark of Cerilliant Corporation