missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Salicylic acid-d4, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001955
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001955 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001955 Supplier Merck Emd Millipore Supplier No. S0421ML

Certified reference material

An internal standard suitable for numerous LC/MS and GC/MS applications from clinical and diagnostic testing to pharmaceutical research, phytochemical testing, and isotope dilution methods. Salicylic acid, the major metabolite of aspirin, is also a phytohormone found in plants with roles in plant growth and development, photosynthesis, transpiration, ion uptake, and transport.

Specifications

Packaging Ampule
CAS 75-05-8, 78646-17-0
Flash Point 2°C (35.6°F) - closed cup
Concentration 100 μg/mL in acetonitrile
Linear Formula C7H2D4O3
MDL Number MFCD06658892
UN Number UN 1648 3 / PGII
Quantity 1 mL
Formula Weight 142.15

CERILLIANT is a registered trademark of Cerilliant Corporation