Learn More
CERILLIANT™ Salicylic acid, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: Merck Emd Millipore S0191ML
Description
Salicylic acid, the major metabolite of aspirin, is also a phytohormone found in plants with roles in plant growth and development, photosynthesis, transpiration, ion uptake, and transport. This certified solution standard is suitable for numerous LC/MS and GC/MS applications including clinical and diagnostic testing, pharmaceutical research and phytochemical testing.Specifications
| Ampule | |
| 0.782 g/cm3 at 20°C | |
| 1.0 mg/mL in acetonitrile | |
| MFCD00002439 | |
| 158°C to 161°C (literature) | |
| 2-Hydroxybenzoic acid | |
| 138.12 |
| 75-05-8,69-72-7 | |
| 2°C (35.6°F) - closed cup | |
| 2-(HO)C6H4CO2H | |
| UN 1648 3 / PGII | |
| 211°C (literature) | |
| 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation