missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ S(-)-Methcathinone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001591
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001591 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001591 Supplier Merck Emd Millipore Supplier No. M0551ML
Only null left

Certified reference material

Known by street names 'Em-kat' and 'Cat', methcathinone is an illicit drug with stimulant and euphoric effects. This Snap-N-Spike™ reference solution is suitable for use in GC/MS or LC/MS applications from urine drug testing and clinical toxicology to forensic analysis.

Specifications

CAS 67-56-1, 66514-93-0
Linear Formula C10H13NO · HCl
MDL Number MFCD00673292
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym S(-)-Methcathinone hydrochloride solution
Formula Weight 199.68
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation