missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ S(+)-Methamphetamine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Description
Methamphetamine is a psychostimulant drug of the phenethylamine and amphetamine classes. Known by the street names 'meth,' 'ice,' and 'glass', this Snap-N-Spike™ reference solution is applicable for use in LC/MS or GC/MS applications including clinical toxicology, sports testing, forensic analysis, or urine drug testing. The dextrorotatory isomer of methamphetamine is sold under the trade name Desoxyn™ for treatment of ADHD and obesity.
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C (49.5°F) - closed cup |
| Concentration | 1.0 mg/mL in methanol |
| Linear Formula | C10H15N |
| MDL Number | MFCD00079140 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Synonym | dextro-Methamphetamine |
| Quantity | 1 mL |
| Formula Weight | 149.23 |
CERILLIANT is a registered trademark of Cerilliant Corporation