missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Retinyl acetate Solution, 100 μg/mL in Ethanol (with 0.1% (w/v) BHT; degassed), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002162
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002162 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002162 Supplier Merck Emd Millipore Supplier No. V0401ML

Certified reference material

Retinyl acetate, a natural preform of vitamin A, is classified as generally recognized as safe in the amounts used to fortify foods with vitamin A and is especially recommended for maternal supplementation during pregnancy.

Specifications

CAS 64-17-5, 127-47-9
Linear Formula C22H32O2
MDL Number MFCD00019413
UN Number UN 1170 - class 3 - PG 2 - Ethanol
Concentration 100 μg/mL in ethanol (with 0.1% (w/v) BHT; degassed)
Synonym Vitamin A acetate solution
Formula Weight 328.49
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation