missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Retinol Solution, 100 μg/mL +/- 25% (Ethanol with 0.1% (w/v) BHT), Sold by MilliporeSigma™ Supelco™

Catalog No. 01002140
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01002140 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01002140 Supplier Merck Emd Millipore Supplier No. V0111ML

Certified reference material

Specifications

CAS 64-17-5, 68-26-8
Linear Formula C20H30O
MDL Number MFCD00001552
UN Number UN 1170 - class 3 - PG 2 - Ethanol, solution
Concentration 100 μg/mL +/- 25% (ethanol with 0.1% (w/v) BHT)
Synonym Retinol solution, 3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexenyl)-2,4,6,8-nonatetraen-1-ol, Axerophthol, Vitamin A alcohol solution, Vitamin A1, all-trans-Retinol
Formula Weight 286.45
Packaging Ampule
Quantity 1 mL
Flash Point 14°C (57.2°F)

CERILLIANT is a registered trademark of Cerilliant Corporation