missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Remifentanil Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001928
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001928 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001928 Supplier Merck Emd Millipore Supplier No. R0241ML

Certified reference material

Remifentanil is a potent, short-acting, synthetic opioid analgesic drug. It is used in combination with other opioids and analgesics to induce or supplement general anaesthesia. This new certified reference solution is suitable for a variety of mass spectrometry-based testing applications including clinical toxicology, forensic analysis, workplace drug testing and pharmaceutical research.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 100 μg/mL in methanol (as free base)
Linear Formula C20H28N2O5 · HCl
MDL Number MFCD00910882
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 412.91

CERILLIANT is a registered trademark of Cerilliant Corporation