missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Remifentanil acid Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001930
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001930 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001930 Supplier Merck Emd Millipore Supplier No. R0261ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material

Remifentanil acid is the major urinary metabolite of remifentanil, a synthetic opioid used to induce or supplement general anesthesia. This new certified reference solution is suitable for a variety of mass spectrometry-based testing applications including clinical toxicology, forensic analysis, workplace drug testing and pharmaceutical research.

Specifications

Packaging Ampule
CAS 75-05-8
Flash Point 2°C (35.6°F) - closed cup
Concentration 100 μg/mL in acetonitrile
Linear Formula C19H26N2O5
UN Number UN 1648 3 / PGII
Quantity 1 mL
Formula Weight 362.42

CERILLIANT is a registered trademark of Cerilliant Corporation