missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ R(-)-Methamphetamine Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001580
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001580 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001580 Supplier Merck Emd Millipore Supplier No. M0241ML
Only null left

Certified reference material

R(-)-Methamphetamine, or levomethamphetamine, is a vasoconstrictor and component of multiple over-the-counter nasal decongestants. This certified Snap-N-Spike™ solution is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as forensic analysis, sports testing, clinical toxicology, or urine drug testing. R(-)-Methamphetamine belongs to the amphetamine and phenethylamine classes of drugs.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol
Linear Formula C10H15N
MDL Number MFCD00135973
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym levo-Methamphetamine
Quantity 1 mL
Formula Weight 149.23

CERILLIANT is a registered trademark of Cerilliant Corporation