missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Quetiapine carboxylic acid-D8 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Quetiapine carboxylic acid is the significant urinary metabolite of quetiapine, marketed as the atypical antipsychotic Seroquel™ for the treatment of depression, bipolar disorder, and schizophrenia. This new internal standard of quetiapine carboxylic acid is applicable to numerous LC/MS and GC/MS applications including quantitation of patient levels, forensic and forensic toxicology analysis, isotope dilution methods, and calibrator preparation.
Specifications
Specifications
| CAS | 75-05-8 |
| Linear Formula | C21H15D8N3O3S |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 405.54 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation