missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Pyrazolam Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001882
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001882 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001882 Supplier Merck Emd Millipore Supplier No. P1181ML

Certified reference material

Pyrazolam is a designer benzodiazepine that has been sold online as ′legal high′ or ′research chemical′. Since its emergence in Europe a few years ago, pyrazolam along with other designer benzodiazepines have increasingly been encountered in forensic case work. This Certified Spiking Solution™ is suitable for a variety of GC/MS or LC/MS testing applications including for use in forensic analysis, urine drug testing, or clinical toxicology.

Specifications

CAS 75-05-8, 39243-02-2
Linear Formula C16H12BrN5
MDL Number MFCD26954579
UN Number UN 1648 3 / PGII
Concentration 1.0 mg/mL in acetonitrile
Formula Weight 354.20
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation