missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Pyrazolam-D4 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 75-05-8 |
| Linear Formula | C16H8D4BrN5 |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 358.23 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 6°C (42.8°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation