missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ (+/-)-Propoxyphene-D5 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Also known as dextropropoxyphene, propoxyphene is an opioid analgesic drug intended for treatment of mild pain. In combination with either paracetamol or aspirin, dextropropoxyphene is sold under trade names including Darvocet-N™ and Darvon™. This Certified Spiking Solution™ is suitable for use as an internal standard in propoxyphene testing methods by GC/MS or LC/MS for applications in urine drug testing, pain prescription monitoring, forensic analysis, or clinical toxicology.
Specifications
Specifications
| CAS | 75-05-8, 136765-49-6 |
| Linear Formula | C22D5H24NO2 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 344.50 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation