missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Progesterone-D9 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Progesterone, a steroid hormone of the progestogen class, is used for hormone replacement therapy. This internal standard is applicable for progesterone testing or isotope dilution methods by GC/MS or LC/MS for endocrinology, female health testing or clinical chemistry.
Specifications
Specifications
| CAS | 75-05-8, 15775-74-3 |
| Linear Formula | C21H21O2D9 |
| MDL Number | MFCD01317491 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 100 μg/mL in acetonitrile |
| Formula Weight | 323.52 |
| Packaging | Ampule |
| Quantity | 1 mL |
CERILLIANT is a registered trademark of Cerilliant Corporation