missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Pregnenolone Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001873
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001873 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001873 Supplier Merck Emd Millipore Supplier No. P1041ML

Certified reference material

Pregnenolone is an endogenous steroid hormone involved in the biological synthesis of other hormones. Pregnenolone is monitored as an ancillary test for congenital adrenal hyperplasia (CAH), particularly in situations where diagnosis of 21-hydroxylase and 11-hydroxylase defficiency has been ruled out. This Certified Spiking Solution™ is suitable for use in LC-MS/MS testing methods as a starting material in the preparation of calibrators, controls and linearity standards.

Specifications

CAS 75-05-8, 145-13-1
Linear Formula C21H32O2
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 316.48
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation