Learn More
CERILLIANT™ Phenytoin Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier:  Merck Emd Millipore P0631ML
Catalog No. 01001843
Only null left
Explore available promotions
Description
Description
Phenytoin is an antiepileptic drug used in the prevention and treatment of seizures. This analytical reference standard is suitable as starting material for the preparation of calibrators and controls in phenytoin testing by GC/MS or LC-MS/MS. Phenytoin is sold under the trade name Dilantin™.
Specifications
Specifications
67-56-1,57-41-0
UN 1230 - class 3 - PG 2 - Methanol, solution
252.27
1 mL
C15H12N2O2
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
CERILLIANT is a registered trademark of Cerilliant Corporation