missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Phenobarbital-D5 (side chain) Solution, (deuterium label on side chain), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001829
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001829 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001829 Supplier Merck Emd Millipore Supplier No. P0171ML
Only null left

Certified reference material

An internal standard suitable for quantitation of phenobarbital levels in GC/MS or LC/MS applications for clinical toxicology, urine drug testing, or forensic analysis. Phenobarbital is a barbiturate used to treat all epileptic seizure types except for absence seizures. Phenobarbital is the oldest and most widely used anticonvulsant.

Specifications

CAS 67-56-1, 73738-05-3
Linear Formula C12D5H7N2O3
MDL Number MFCD00145091
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Formula Weight 237.27
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation